Assay Details
Target Gene Details
Entrez Gene ID: | 15422 |
Gene Name: | homeobox C13 |
Gene Aliases: |
- |
Location: |
Chr.15:102921116-102929135 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hoxc13 | NM_010464.2 | 1 | 263 | NP_034594.1 |
| AF193796.1 | 1 | 263 | AAL09298.1 | |
| AY349616.1 | 1 | 291 | AAQ54690.1 |

Genomic Map