Assay Details
Target Gene Details
Entrez Gene ID: | 17309 |
Gene Name: | mannoside acetylglucosaminyltransferase 3 |
Gene Aliases: |
1110038J12Rik, AI848272, GnT-III |
Location: |
Chr.15:80173650-80215519 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mgat3 | NM_010795.4 | 2 | 1637 | NP_034925.2 |
| AK081850.1 | 2 | 1701 | ||
| BC053040.1 | 2 | 1547 | AAH53040.1 | |
| BC059828.1 | 1 | 1529 |

Genomic Map