Assay Details
Target Gene Details
Entrez Gene ID: | 72555 |
Gene Name: | shisa family member 9 |
Gene Aliases: |
2700045P11Rik, Ckamp44 |
Location: |
Chr.16:11984113-12270904 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Shisa9 | NM_001174086.1 | 4 | 3053 | NP_001167557.1 |
| NM_028277.2 | 5 | 3101 | NP_082553.2 | |
| AK049860.1 | 5 | 2627 |

Genomic Map