Assay Details
Target Gene Details
Entrez Gene ID: | 66598 |
Gene Name: | RIKEN cDNA 3110001I22 gene |
Gene Aliases: |
- |
Location: |
Chr.16:13672020-13678385 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 3110001I22Rik | NM_025653.2 | 2 | 826 | NP_079929.1 |
| AK012939.1 | 2 | 827 | BAB28561.1 | |
| AK013950.1 | 2 | 897 | ||
| AK031567.1 | 2 | 860 | BAC27453.1 | |
| BC038880.1 | 1 | 645 | AAH38880.1 |
Target Gene Details
Entrez Gene ID: | 67118 |
Gene Name: | bifunctional apoptosis regulator |
Gene Aliases: |
3010001A07Rik, AI666707, AW107665, Bar, Rnf47 |
Location: |
Chr.16:13671858-13703612 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bfar | NM_001177552.1 | NP_001171023.1 | ||
| NM_025976.5 | NP_080252.1 | |||
| AK013874.1 | BAB29029.1 | |||
| AK028592.1 | BAC26022.1 | |||
| AK041839.1 | ||||
| BC027221.1 | AAH27221.1 | |||
| BY329099.1 |
More Information
Set Membership: |
|

Genomic Map