Assay Details
Target Gene Details
Entrez Gene ID: | 18996 |
Gene Name: | POU domain, class 4, transcription factor 1 |
Gene Aliases: |
Brn-3, Brn-3.0, Brn3, Brn3.0, Brn3a, E130119J07Rik |
Location: |
Chr.14:104461675-104468022 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pou4f1 | NM_011143.4 | 1 | 269 | NP_035273.3 |
| AY706205.1 | 1 | 269 | AAU13951.1 |

Genomic Map