Assay Details
Target Gene Details
Entrez Gene ID: | 223921 |
Gene Name: | achalasia, adrenocortical insufficiency, alacrimia |
Gene Aliases: |
AAA, ADRACALA, D030041N15Rik, GL003 |
Location: |
Chr.15:102338247-102350759 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aaas | NM_153416.2 | 16 | 1643 | NP_700465.2 |
| AK034591.1 | 16 | 1538 | BAC28764.1 | |
| AK079849.1 | 15 | 1651 | ||
| AK083401.1 | 16 | 1557 | ||
| AK083537.1 | 16 | 1564 | BAC38945.1 | |
| AK087134.1 | 16 | 1619 | BAC39811.1 | |
| AK088247.1 | 16 | 1560 | BAC40234.1 | |
| AK134406.1 | 16 | 1534 | BAE22131.1 | |
| AK165243.1 | 16 | 1558 | BAE38099.1 | |
| AK166118.1 | 16 | 1558 | BAE38581.1 | |
| AK169523.1 | 16 | 1595 | BAE41208.1 | |
| BB397636.1 | 1 | 118 | ||
| BC018191.1 | 16 | 1544 | AAH18191.1 | |
| BC025501.1 | 16 | 1590 | AAH25501.1 |

Genomic Map