Assay Details
Target Gene Details
Entrez Gene ID: | 66270 |
Gene Name: | family with sequence similarity 134, member B |
Gene Aliases: |
1810015C04Rik, AU015349 |
Location: |
Chr.15:25843298-25973696 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam134b | NM_001034851.2 | 9 | 1397 | NP_001030023.1 |
| NM_001277315.1 | 7 | 1486 | NP_001264244.1 | |
| NM_001277316.1 | 7 | 1069 | NP_001264245.1 | |
| NM_001277317.1 | 7 | 1047 | NP_001264246.1 | |
| NM_001277318.1 | 7 | 1019 | NP_001264247.1 | |
| NM_025459.3 | 8 | 1187 | NP_079735.2 | |
| AK007506.1 | 7 | 1048 | BAB25077.1 | |
| AK015182.1 | 8 | 1009 | ||
| AK015887.1 | 1 | 366 | BAB30019.1 | |
| AK146972.1 | 7 | 1070 | BAE27577.1 | |
| AK167327.1 | 7 | 1487 | BAE39429.1 | |
| BC019494.1 | 8 | 1126 | AAH19494.1 | |
| BC054415.1 | 7 | 1006 | AAH54415.1 |

Genomic Map