Assay Details
Target Gene Details
Entrez Gene ID: | 223601 |
Gene Name: | family with sequence similarity 49, member B |
Gene Aliases: |
0910001A06Rik, AW122079 |
Location: |
Chr.15:63929094-64060452 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam49b | NM_144846.5 | 12 | 3373 | NP_659095.1 |
| AK028062.1 | 12 | 3374 | BAC25731.1 | |
| BX527479.1 | 1 | 113 |

Genomic Map