Assay Details
Target Gene Details
Entrez Gene ID: | 19082 |
Gene Name: | protein kinase, AMP-activated, gamma 1 non-catalytic subunit |
Gene Aliases: |
AA571379, BB036179, Prkaac |
Location: |
Chr.15:98812797-98831508 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prkag1 | NM_016781.2 | 12 | 1189 | NP_058061.2 |
| AF036535.1 | 12 | 1161 | AAB95475.1 | |
| AK138324.1 | 12 | 1155 | BAE23624.1 | |
| AK159583.1 | 12 | 1133 | BAE35203.1 | |
| BC086660.1 | 12 | 1135 | AAH86660.1 |
More Information
Set Membership: |
|

Genomic Map