Assay Details
Target Gene Details
Entrez Gene ID: | 17695 |
Gene Name: | beta-microseminoprotein |
Gene Aliases: |
PIP, PSP94, beta-MSP |
Location: |
Chr.14:32142023-32158327 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Msmb | NM_020597.3 | 4 | 308 | NP_065622.1 |
| U89840.1 | 3 | 276 | AAB49683.1 |
More Information
Set Membership: |
|

Genomic Map