Assay Details
Target Gene Details
Entrez Gene ID: | 239337 |
Gene Name: | a disintegrin-like and metallopeptidase (reprolysin type) with thrombospondin type 1 motif, 12 |
Gene Aliases: |
ADAMTS-12, AI605170 |
Location: |
Chr.15:11064790-11346867 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adamts12 | NM_175501.2 | 1 | 246 | NP_780710.2 |
| AJ537452.1 | CAD60967.1 | |||
| AK048612.1 | 1 | 209 | BAC33391.1 | |
| AK054015.1 | 1 | 218 | BAC35621.1 | |
| AK135354.1 | 1 | 248 | BAE22500.1 | |
| AK145096.1 | 1 | 348 | BAE26232.1 |
More Information
Set Membership: |
|

Genomic Map