Assay Details
Target Gene Details
Entrez Gene ID: | 106059 |
Gene Name: | RIKEN cDNA A430088P11 gene |
Gene Aliases: |
- |
Location: |
Chr.15:80691025-80697610 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A430088P11Rik | NR_045309.1 | 4 | 3212 | |
| NR_045310.1 | 3 | 3126 | ||
| AK040361.1 | 3 | 3126 |
Target Gene Details
Entrez Gene ID: | 213956 |
Gene Name: | family with sequence similarity 83, member F |
Gene Aliases: |
AW544981 |
Location: |
Chr.15:80671847-80700425 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam83f | NM_145986.2 | 4 | 1176 | NP_666098.2 |
| AK030342.1 | 4 | 1158 | BAC26912.1 | |
| AK145865.1 | 4 | 1176 | BAE26707.1 | |
| BC016600.1 | 4 | 1154 |

Genomic Map