Assay Details
Target Gene Details
Entrez Gene ID: | 21357 |
Gene Name: | TAR (HIV) RNA binding protein 2 |
Gene Aliases: |
Prbp, TRBP |
Location: |
Chr.15:102518192-102523676 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tarbp2 | NM_001253795.1 | 1 | 233 | NP_001240724.1 |
| NM_009319.3 | 1 | 233 | NP_033345.2 | |
| AK143188.1 | 1 | 233 | ||
| AK144205.1 | 1 | 71 | BAE25768.1 | |
| AV582078.1 | 1 | 91 | ||
| BI737851.1 | 1 | 179 |

Genomic Map