Assay Details
Target Gene Details
Entrez Gene ID: | 15209 |
Gene Name: | homeobox gene expressed in ES cells |
Gene Aliases: |
HES-1, Rpx |
Location: |
Chr.14:26994420-27002329 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hesx1 | NM_010420.2 | 1 | 78 | NP_034550.2 |
| U40720.1 | 1 | 78 | AAA97869.1 |
More Information
Set Membership: |
|

Genomic Map