Assay Details
Target Gene Details
Entrez Gene ID: | 74385 |
Gene Name: | adaptor-related protein complex 5, mu 1 subunit |
Gene Aliases: |
4932432K03Rik, Mu5, Mudeng |
Location: |
Chr.14:49066495-49094114 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ap5m1 | NM_144535.4 | 1 | 123 | NP_653118.3 |
| AK016540.1 | 1 | 70 | ||
| AK028927.1 | 1 | 76 | ||
| AK030928.1 | BAC27187.1 | |||
| AK031527.1 | 1 | 522 | BAC27435.1 | |
| AK036454.1 | ||||
| AK036465.1 | 1 | 55 | BAC29441.1 | |
| AK041400.1 | 1 | 76 | ||
| AK042544.1 | BAC31288.2 | |||
| AK051372.1 | 1 | 70 | BAC34616.1 | |
| AK154166.1 | 1 | 76 | BAE32417.1 | |
| BC023094.1 | 1 | 123 | AAH23094.1 |
Target Gene Details
Entrez Gene ID: | 105504 |
Gene Name: | exocyst complex component 5 |
Gene Aliases: |
AI447711, AI448003, Gm76, PRO1912, SEC10, Sec10l1 |
Location: |
Chr.14:49011922-49066667 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Exoc5 | NM_207214.3 | 1 | 51 | NP_997097.1 |
More Information
Set Membership: |
|

Genomic Map