Assay Details
Target Gene Details
Entrez Gene ID: | 268822 |
Gene Name: | aarF domain containing kinase 5 |
Gene Aliases: |
A230108P17 |
Location: |
Chr.15:76576359-76595812 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adck5 | NM_172960.3 | 13 | 1557 | NP_766548.2 |
| NR_073123.1 | 13 | 1552 | ||
| AK039217.1 | 13 | 1553 | BAC30280.1 | |
| AK163557.1 | 12 | 1584 | ||
| BC060528.1 | 13 | 1441 | AAH60528.1 |
More Information
Set Membership: |
|

Genomic Map