Assay Details
Target Gene Details
Entrez Gene ID: | 223604 |
Gene Name: | potassium channel, subfamily K, member 9 |
Gene Aliases: |
Task3 |
Location: |
Chr.15:72502927-72547853 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnk9 | NM_001033876.1 | 2 | 439 | NP_001029048.1 |
| BC139167.1 | 2 | 500 | ||
| BC139168.1 | 2 | 500 | ||
| DQ185133.1 | 2 | 439 | ABA28314.1 |
More Information
Set Membership: |
|

Genomic Map