Assay Details
Target Gene Details
Entrez Gene ID: | 67498 |
Gene Name: | potassium channel, subfamily V, member 1 |
Gene Aliases: |
2700023A03Rik, vibe |
Location: |
Chr.15:45106237-45116488 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcnv1 | NM_026200.3 | 3 | 2367 | NP_080476.2 |
| AK012275.1 | 3 | 2162 | BAB28134.1 | |
| AK034091.1 | 3 | 2260 | BAC28581.1 | |
| AK139482.1 | 3 | 2203 | BAE24032.1 |
More Information
Set Membership: |
|

Genomic Map