Assay Details
Target Gene Details
Entrez Gene ID: | 17869 |
Gene Name: | myelocytomatosis oncogene |
Gene Aliases: |
AU016757, Myc2, Niard, Nird, bHLHe39 |
Location: |
Chr.15:61985341-61990361 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Myc | NM_001177352.1 | 2 | 1249 | NP_001170823.1 |
| NM_001177353.1 | 2 | 1246 | NP_001170824.1 | |
| NM_001177354.1 | 2 | 1246 | NP_001170825.1 | |
| NM_010849.4 | 2 | 1249 | NP_034979.3 | |
| AF076523.1 | AAC26879.1 | |||
| AK087961.1 | 2 | 1041 | BAC40060.1 | |
| AK133952.1 | 2 | 1023 | BAE21948.1 | |
| AK145084.1 | 2 | 1203 | BAE26228.1 | |
| BC006728.1 | 1 | 723 | ||
| BC138931.1 | 2 | 712 | ||
| BC138932.1 | 2 | 712 | ||
| X01023.1 | 2 | 1229 | CAA25508.1 |
More Information
Set Membership: |
|

Genomic Map