Assay Details
Target Gene Details
Entrez Gene ID: | 27406 |
Gene Name: | ATP-binding cassette, sub-family F (GCN20), member 3 |
Gene Aliases: |
AI326318, AU016058, BB119416 |
Location: |
Chr.16:20548556-20561603 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcf3 | NM_013852.2 | 7 | 684 | NP_038880.1 |
| AK030626.1 | 7 | 648 | ||
| AK183731.1 | ||||
| AK189060.1 | ||||
| AK214630.1 | 4 | 315 | ||
| BC032923.1 | 7 | 625 | AAH32923.1 |
More Information
Set Membership: |
|

Genomic Map