Assay Details
Target Gene Details
Entrez Gene ID: | 224405 |
Gene Name: | cysteine and tyrosine-rich protein 1 |
Gene Aliases: |
- |
Location: |
Chr.16:85450357-85550905 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyyr1 | NM_144853.3 | 1 | 170 | NP_659102.1 |
| AK031840.1 | 1 | 193 | ||
| AK158088.1 | 1 | 228 | ||
| AK171936.1 | 1 | 195 | ||
| AY061854.1 | 1 | 192 | AAL35738.1 | |
| BC099957.1 | 1 | 170 | AAH99957.1 | |
| BC125594.1 | ||||
| BC125596.1 |

Genomic Map