Assay Details
Target Gene Details
Entrez Gene ID: | 15209 |
Gene Name: | homeobox gene expressed in ES cells |
Gene Aliases: |
HES-1, Rpx |
Location: |
Chr.14:26994420-27002329 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hesx1 | NM_010420.2 | 4 | 862 | NP_034550.2 |
| AK082831.1 | 4 | 510 | BAC38643.1 | |
| BC132051.1 | 4 | 688 | ||
| BC132053.1 | 4 | 688 | ||
| BC145262.1 | 4 | 687 | ||
| U40720.1 | 4 | 860 | AAA97869.1 | |
| X80040.1 | 4 | 570 | CAA56344.1 |
More Information
Set Membership: |
|

Genomic Map