Assay Details
Target Gene Details
Entrez Gene ID: | 56187 |
Gene Name: | Rab geranylgeranyl transferase, a subunit |
Gene Aliases: |
gm |
Location: |
Chr.14:55715877-55722368 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabggta | NM_019519.2 | 16 | 2108 | NP_062392.1 |
| AF127656.1 | 17 | 1956 | AAF65920.1 | |
| AF127657.1 | 16 | 1899 | ||
| AF127658.1 | 16 | 2108 | AAF65921.1 | |
| AF127659.1 | 16 | 2108 | AAF65922.1 | |
| AF127660.1 | 16 | 2053 | AAF65923.1 | |
| AF127661.1 | 15 | 1996 | ||
| AF127662.1 | 17 | 2027 | AAF65924.1 | |
| AK002625.1 | BAB22240.1 | |||
| AK146917.1 | 17 | 1979 | BAE27529.1 | |
| BC012214.1 | AAH12214.1 |
More Information
Set Membership: |
|

Genomic Map