Assay Details
Target Gene Details
Entrez Gene ID: | 100502987 |
Gene Name: | predicted gene 17066 |
Gene Aliases: |
- |
Location: |
Chr.14:105106728-105114676 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm17066 | NR_040628.1 | |||
| AK005802.1 |
Target Gene Details
Entrez Gene ID: | 74213 |
Gene Name: | RNA binding motif protein 26 |
Gene Aliases: |
1700009P03Rik, AI447318, C230097K14Rik, C86225, MLZ-393, Pro1777, Se70-2 |
Location: |
Chr.14:105114519-105177327 on Build GRCm38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbm26 | NM_134077.4 | 21 | 3977 | NP_598838.3 |
| BC029079.1 | 8 | 1423 | AAH29079.1 |
More Information
Set Membership: |
|

Genomic Map