Assay Details
Target Gene Details
Entrez Gene ID: | 19661 |
Gene Name: | retinol binding protein 3, interstitial |
Gene Aliases: |
Irbp, Rbp-3 |
Location: |
Chr.14:33954022-33964216 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rbp3 | NM_015745.2 | 1 | 1048 | NP_056560.1 |
| BC017610.1 | 1 | 1048 | AAH17610.1 | |
| BC042480.1 | 1 | 1010 | AAH42480.1 | |
| BC083133.1 | 1 | 1067 | AAH83133.1 |

Genomic Map