Assay Details
Target Gene Details
Entrez Gene ID: | 52589 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
AI848120, D030020D09Rik, D15Ertd412e |
Location: |
Chr.15:37366175-37792519 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncald | NM_134094.4 | 1 | 110 | NP_598855.2 |
| AK048440.1 | 1 | 90 | BAC33338.1 |

Genomic Map