Assay Details
Target Gene Details
Entrez Gene ID: | 52589 |
Gene Name: | neurocalcin delta |
Gene Aliases: |
AI848120, D030020D09Rik, D15Ertd412e |
Location: |
Chr.15:37366175-37792519 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ncald | NM_001170866.1 | 7 | 3081 | NP_001164337.1 |
| NM_001170867.1 | 5 | 3015 | NP_001164338.1 | |
| NM_001170868.1 | 4 | 2963 | NP_001164339.1 | |
| NM_134094.4 | 7 | 3271 | NP_598855.2 | |
| AK034031.1 | 5 | 2994 | BAC28553.1 | |
| AK048440.1 | 7 | 3251 | BAC33338.1 | |
| AK085416.1 | 1 | 1619 | ||
| AK167313.1 | 4 | 2933 | BAE39416.1 | |
| BC011162.1 | 4 | 2958 | AAH11162.1 | |
| BC026979.1 | 4 | 2917 | AAH26979.1 |

Genomic Map