Assay Details
Target Gene Details
Entrez Gene ID: | 73379 |
Gene Name: | discoidin, CUB and LCCL domain containing 2 |
Gene Aliases: |
1700055P21Rik, AW146002, CLCP1, Esdn |
Location: |
Chr.16:58408426-58469745 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcbld2 | NM_028523.3 | 1 | 65 | NP_082799.2 |
| AF387548.1 | 1 | 157 | AAL30179.1 | |
| AF442820.1 | 1 | 179 | AAL30434.1 | |
| BC066097.1 | 1 | 65 | AAH66097.1 |

Genomic Map