Assay Details
Target Gene Details
Entrez Gene ID: | 73379 |
Gene Name: | discoidin, CUB and LCCL domain containing 2 |
Gene Aliases: |
1700055P21Rik, AW146002, CLCP1, Esdn |
Location: |
Chr.16:58408426-58469745 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcbld2 | NM_028523.3 | 16 | 4266 | NP_082799.2 |
| AK043915.1 | 1 | 55 | ||
| AK051889.1 | 14 | 3544 | BAC34801.1 | |
| BC066097.1 | 16 | 4260 | AAH66097.1 |

Genomic Map