Assay Details
Target Gene Details
Entrez Gene ID: | 58239 |
Gene Name: | dexamethasone-induced transcript |
Gene Aliases: |
1810029J14Rik, AI836170, AW413143, D16Bwg0586e, Myle |
Location: |
Chr.16:10530207-10543122 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dexi | NM_021428.4 | 2 | 1091 | NP_067403.2 |
| AF152470.2 | 2 | 792 | AAD34163.2 | |
| AK007644.1 | 2 | 1092 | BAB25159.2 | |
| BC017551.1 | 2 | 1078 | AAH17551.1 |

Genomic Map