Assay Details
Target Gene Details
Entrez Gene ID: | 13356 |
Gene Name: | DiGeorge syndrome critical region gene 2 |
Gene Aliases: |
9930034O06Rik, DGS-C, Dgsc, Idd, Lan, Sez12, mKIAA0163 |
Location: |
Chr.16:17840355-17894098 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dgcr2 | NM_001109750.1 | 10 | 2459 | NP_001103220.1 |
| NM_010048.3 | 10 | 2468 | NP_034178.2 | |
| AK088131.1 | 9 | 2185 | BAC40164.1 | |
| AK129070.1 | 11 | 2318 | BAC97880.1 | |
| AK133880.1 | 7 | 1757 | BAE21909.1 | |
| AK141594.1 | 10 | 2434 | BAE24753.1 | |
| AK145495.1 | 10 | 2385 | BAE26471.1 | |
| AK153026.1 | 10 | 2255 | ||
| AK163872.1 | 10 | 2421 | BAE37523.1 | |
| AK163903.1 | 10 | 2397 | ||
| AK164263.1 | 3 | 1168 | BAE37709.1 | |
| AK183515.1 | 1 | 84 | ||
| BC031506.1 | 3 | 1178 | AAH31506.1 | |
| BC060636.1 | 10 | 2409 | AAH60636.1 | |
| BC062978.1 | 10 | 2363 | AAH62978.1 | |
| D78641.1 | 10 | 2393 | BAA11460.1 | |
| X95480.1 | 10 | 2334 | CAA64749.1 |

Genomic Map