Assay Details
Target Gene Details
Entrez Gene ID: | 140743 |
Gene Name: | rad and gem related GTP binding protein 2 |
Gene Aliases: |
AW411893 |
Location: |
Chr.14:54476100-54480434 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rem2 | NM_080726.3 | 1 | 47 | NP_542764.2 |
| AK090165.1 | 1 | 48 | BAC41120.1 |

Genomic Map