Assay Details
Target Gene Details
Entrez Gene ID: | 67434 |
Gene Name: | ankyrin repeat domain 33B |
Gene Aliases: |
0610012A05Rik, 3021401C12Rik, 5730557B15Rik |
Location: |
Chr.15:31291227-31368894 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd33b | NM_001164441.1 | 1 | 531 | NP_001157913.1 |
| NM_026153.3 | 1 | 531 | NP_080429.1 | |
| NM_027496.3 | 1 | 531 | NP_081772.1 | |
| AK002572.1 | 1 | 326 | BAB22197.1 | |
| AK012871.1 | 1 | 341 | BAB28525.1 | |
| AK017844.1 | 1 | 337 | BAB30970.1 | |
| AK089667.1 | 1 | 305 | BAC40941.1 | |
| AK156757.1 | 1 | 499 | BAE33840.1 | |
| AK170894.1 | 1 | 305 | BAE42099.1 |

Genomic Map