Assay Details
Target Gene Details
Entrez Gene ID: | 67434 |
Gene Name: | ankyrin repeat domain 33B |
Gene Aliases: |
0610012A05Rik, 3021401C12Rik, 5730557B15Rik |
Location: |
Chr.15:31291227-31368894 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankrd33b | NM_001164441.1 | 4 | 1872 | NP_001157913.1 |
| NM_027496.3 | 4 | 1845 | NP_081772.1 | |
| AK017844.1 | 4 | 1650 | BAB30970.1 | |
| AK089667.1 | 5 | 1743 | BAC40941.1 | |
| AK156757.1 | 4 | 1840 | BAE33840.1 |

Genomic Map