Assay Details
Target Gene Details
Entrez Gene ID: | 106039 |
Gene Name: | golgi associated, gamma adaptin ear containing, ARF binding protein 1 |
Gene Aliases: |
4930406E12Rik, AU016030, AW209092 |
Location: |
Chr.15:78877190-78894585 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gga1 | NM_145929.2 | 17 | 2750 | NP_666041.1 |
| AK080881.1 | 17 | 2373 | BAC38058.1 | |
| AK155195.1 | 17 | 2390 | BAE33109.1 | |
| AK159771.1 | 17 | 2373 | ||
| AK170437.1 | 17 | 2389 | ||
| BC026802.1 | 17 | 2750 | AAH26802.1 |

Genomic Map