Assay Details
Target Gene Details
Entrez Gene ID: | 246133 |
Gene Name: | potassium voltage-gated channel, Isk-related subfamily, gene 2 |
Gene Aliases: |
2200002I16Rik, AW048273, MiRP1 |
Location: |
Chr.16:92292389-92298137 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcne2 | NM_134110.3 | 2 | 392 | NP_598871.1 |
| AK008619.1 | 2 | 393 | BAB25781.1 | |
| BC022699.1 | 2 | 334 | AAH22699.1 |
More Information
Set Membership: |
|

Genomic Map