Assay Details
Target Gene Details
Entrez Gene ID: | 16391 |
Gene Name: | interferon regulatory factor 9 |
Gene Aliases: |
Irf-9, Isgf3g, p48 |
Location: |
Chr.14:55603985-55610030 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Irf9 | NM_001159417.1 | 1 | 124 | NP_001152889.1 |
| NM_008394.3 | 1 | 124 | NP_032420.1 | |
| CN531811.1 | 1 | 538 |
More Information
Set Membership: |
|

Genomic Map