Assay Details
Target Gene Details
Entrez Gene ID: | 16424 |
Gene Name: | inter-alpha trypsin inhibitor, heavy chain 1 |
Gene Aliases: |
ITI-HC1, Intin1, Itih-1 |
Location: |
Chr.14:30929180-30943321 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Itih1 | NM_008406.3 | 22 | 2769 | NP_032432.2 |
| AK149500.1 | 22 | 2770 | BAE28922.1 | |
| BC013465.1 | 22 | 2753 | AAH13465.1 | |
| BC028814.1 | 22 | 2769 | AAH28814.1 | |
| X70391.1 | 22 | 2749 | CAA49841.1 |
More Information
Set Membership: |
|

Genomic Map