Assay Details
Target Gene Details
Entrez Gene ID: | 497113 |
Gene Name: | ribonuclease, RNase A family, 11 (non-active) |
Gene Aliases: |
Raj1 |
Location: |
Chr.14:51048694-51055702 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase11 | NM_001011877.2 | 1 | 271 | NP_001011877.1 |
| BC141252.1 | 1 | 271 |

Genomic Map