Assay Details
Target Gene Details
Entrez Gene ID: | 497106 |
Gene Name: | ribonuclease, RNase A family, 12 (non-active) |
Gene Aliases: |
Rai1 |
Location: |
Chr.14:51056698-51057242 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase12 | NM_001011875.2 | 1 | 247 | NP_001011875.1 |
| BC146014.1 | 1 | 247 | ||
| BC146016.1 | 1 | 247 |

Genomic Map