Assay Details
Target Gene Details
Entrez Gene ID: | 11727 |
Gene Name: | angiogenin, ribonuclease, RNase A family, 5 |
Gene Aliases: |
AI385586, Ang1, Rnase5, Rnase5a |
Location: |
Chr.14:51091077-51102009 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ang | NM_001161731.2 | NP_001155203.1 | ||
| NM_007447.3 | 1 | 161 | NP_031473.1 | |
| AK159966.1 | 1 | 162 | BAE35521.1 | |
| AK171251.1 | BAE42343.1 | |||
| BF721354.1 |
Target Gene Details
Entrez Gene ID: | 58809 |
Gene Name: | ribonuclease, RNase A family 4 |
Gene Aliases: |
C730049F20Rik, Rab1 |
Location: |
Chr.14:51091077-51106151 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase4 | NM_021472.4 | NP_067447.2 | ||
| NM_201239.3 | 1 | 161 | NP_957691.1 | |
| AB041044.1 | BAA96476.1 | |||
| AK076504.1 | BAC36368.1 | |||
| BC005569.1 | AAH05569.1 | |||
| BC061105.1 | AAH61105.1 | |||
| BC066843.1 | AAH66843.1 |

Genomic Map