Assay Details
Target Gene Details
Entrez Gene ID: | 58809 |
Gene Name: | ribonuclease, RNase A family 4 |
Gene Aliases: |
C730049F20Rik, Rab1 |
Location: |
Chr.14:51091077-51106151 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase4 | NM_021472.4 | 2 | 400 | NP_067447.2 |
| NM_201239.3 | 2 | 697 | NP_957691.1 | |
| AB041044.1 | 2 | 306 | BAA96476.1 | |
| AK050451.1 | 2 | 297 | BAC34263.1 | |
| AK076504.1 | 2 | 370 | BAC36368.1 | |
| AY762362.1 | 2 | 299 | AAV31936.1 | |
| BC005569.1 | 2 | 274 | AAH05569.1 | |
| BC061105.1 | 2 | 281 | AAH61105.1 | |
| BC066843.1 | 2 | 361 | AAH66843.1 |

Genomic Map