Assay Details
Target Gene Details
Entrez Gene ID: | 78416 |
Gene Name: | ribonuclease, RNase A family, 6 |
Gene Aliases: |
9530043P15Rik, BB119466 |
Location: |
Chr.14:51123913-51132187 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase6 | NM_030098.2 | 2 | 525 | NP_084374.2 |
| AK020595.1 | 2 | 469 | BAB32143.1 | |
| AK038708.1 | 3 | 841 | BAE20540.1 | |
| AY545655.1 | 2 | 503 | AAS48640.1 | |
| BC094892.1 | 2 | 479 | AAH94892.1 | |
| BC132632.1 | 2 | 458 | ||
| BC132634.1 | 2 | 457 |

Genomic Map