Assay Details
Target Gene Details
Entrez Gene ID: | 328401 |
Gene Name: | ribonuclease, RNase A family, 9 (non-active) |
Gene Aliases: |
ESRL, Rak1 |
Location: |
Chr.14:51038459-51041905 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnase9 | NM_183032.2 | 2 | 840 | NP_898853.2 |
| AK078986.1 | 2 | 843 | BAC37494.1 | |
| AY226989.1 | 2 | 841 | AAP43946.1 | |
| AY303816.1 | 2 | 840 | AAQ73516.1 | |
| BC107218.1 | 2 | 843 | AAI07219.1 |

Genomic Map