Assay Details
Target Gene Details
Entrez Gene ID: | 54151 |
Gene Name: | cysteine and histidine rich 1 |
Gene Aliases: |
1110031M01Rik, AU042374, Chrp |
Location: |
Chr.15:76643395-76660208 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyhr1 | NM_019396.3 | 5 | 1595 | NP_062269.1 |
| AJ251516.1 | 5 | 1448 | CAB62575.1 | |
| AK028994.1 | 1 | 354 | BAC26231.1 | |
| CF740409.1 | 1 | 384 |

Genomic Map