Assay Details
Target Gene Details
Entrez Gene ID: | 54151 |
Gene Name: | cysteine and histidine rich 1 |
Gene Aliases: |
1110031M01Rik, AU042374, Chrp |
Location: |
Chr.15:76643395-76660208 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyhr1 | NM_001276322.1 | 3 | 847 | NP_001263251.1 |
| NM_001276333.1 | 2 | 890 | NP_001263262.1 | |
| NM_001276334.1 | 3 | 674 | NP_001263263.1 | |
| NM_001276335.1 | 3 | 620 | NP_001263264.1 | |
| NM_019396.3 | NP_062269.1 | |||
| NM_180962.2 | 3 | 913 | NP_851293.1 | |
| AJ251516.1 | CAB62575.1 | |||
| AK004009.1 | 3 | 757 | BAB23122.1 | |
| BC016196.1 | 3 | 711 | AAH16196.1 | |
| BC025112.1 | 3 | 813 | AAH25112.1 |

Genomic Map