Assay Details
Target Gene Details
Entrez Gene ID: | 110115 |
Gene Name: | cytochrome P450, family 11, subfamily b, polypeptide 1 |
Gene Aliases: |
AA389061, CPN1, Cyp11b, Cyp11b-1, FHI |
Location: |
Chr.15:74834892-74841643 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cyp11b1 | NM_001033229.3 | 9 | 2072 | NP_001028401.2 |
| AK142439.1 | 9 | 2049 | BAE25066.1 | |
| AK163083.1 | 9 | 2045 | BAE37185.1 |
Target Gene Details
Entrez Gene ID: | 15202 |
Gene Name: | hematopoietic cell transcript 1 |
Gene Aliases: |
1700057K19Rik, HemT |
Location: |
Chr.15:74818880-74846164 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hemt1 |

Genomic Map