Assay Details
Target Gene Details
Entrez Gene ID: | 16687 |
Gene Name: | keratin 6A |
Gene Aliases: |
2310016L08Rik, Krt2-6a, Krt2-6c, MK6a, mK6[a] |
Location: |
Chr.15:101689928-101694305 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Krt6a | NM_008476.3 | 9 | 1869 | NP_032502.3 |
| AK009394.1 | 9 | 1872 | ||
| AK010039.1 | 4 | 762 | BAB26659.1 | |
| BC080820.1 | 9 | 1869 | AAH80820.1 |

Genomic Map