Assay Details
Target Gene Details
Entrez Gene ID: | 67841 |
Gene Name: | autophagy related 3 |
Gene Aliases: |
2610016C12Rik, APG3, Apg3l, Atg3l, PC3-96 |
Location: |
Chr.16:45158829-45188538 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atg3 | NM_026402.3 | 12 | 1570 | NP_080678.1 |
| AK011409.1 | 12 | 1571 | BAB27600.1 |
More Information
Set Membership: |
|

Genomic Map