Assay Details
Target Gene Details
Entrez Gene ID: | 246738 |
Gene Name: | DnaJ (Hsp40) homolog, subfamily C, member 28 |
Gene Aliases: |
BC020175, ORF28 |
Location: |
Chr.16:91614257-91618999 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnajc28 | NM_001099738.1 | 2 | 3019 | NP_001093208.1 |
| NM_138664.2 | 2 | 2849 | NP_619605.2 | |
| AV328433.2 | 1 | 108 | ||
| BC005604.1 | 1 | 1921 | AAH05604.1 |
More Information
Set Membership: |
|

Genomic Map